missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ EA-50 Stain

Product Code. p-4530157 Shop All Epredia Products
Click to view available options
Quantity:
1L
4L
Unit Size:
4 L
Pack of 2
This item is not returnable. View return policy

Product Code. 10167988

Brand: Epredia™ 6766001

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Clearly differentiate cellular components with pink, green and blue-green hues utilizing Epredia™ Shandon™ EA-50 Stain.

  • Polychromatic cytoplasmic stain
  • Biological stain commission-certified dye
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1L
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cytological stain EA-50 / EA-65
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Ethyl alcohol, Methyl alcohol, Isopropylalcohol, Phosphotungstic acid, Eosin-YDye
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.