missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ Eosin-Y Stain
Produce brilliant staining with Epredia™ Shandon™ Eosin-Y, a fast-acting cytoplasmic counterstain.
Brand: Epredia™ 6766009
128.35 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Ready-to-use
- Produces consistent results
- Stable and precipitate-free; no filtering
- Compatible with all alum hematoxylin stains
- Available in alcoholic or aqueous formulations
Specifications
Aqueous Eosin Y Stain | |
1L |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H335-May cause respiratory irritation.
Precautionary Statement
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P370+P378-In case of fire: Use to extinguish.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : formaldehyde, aceticacid, sodium acetate, eosin-Y dye