missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ EZ Single Cytofunnel™ with White Filter Cards and Caps
Optimize cell yield with the Epredia™ EZ Single Cytofunnel™ with White Filter Cards and Caps.
Brand: Epredia™ A78710003
105.61 GBP valid until 2024-12-31
Use promo code "21930" to get your promotional price.
This item is not returnable.
View return policy
Description
- Reduces the risk of exposure to pathogenic samples
- Disposable to eliminate time-consuming decontamination and cleaning
- Decreases the possibility of cross-contamination
- Eliminates the need for metal Cytoclips™
- Produces more cost-effective, thin-layer preparations when compared to other thin-layer methods
- Holds sample volumes up to 0.5 mL
- Includes cell deposition area of 6 mm in diameter
Includes:
- Closure caps
- Easy-to-use operational illustrations
Specifications
White | |
40 Pk. | |
0.1 to 0.5 mL |
Single-use | |
EZ Single Cytofunnel™, With Filter Cards and Caps |