missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Formal-Fixx™ 10% Neutral Buffered Formalin

Produce consistent results and extend life of rotary valve in tissue processors with Epredia™ Formal-Fixx™ 10% Neutral Buffered Foramalin.

£40.65 - £98.10


Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
12673026
View Documents
Epredia™
6764240
5 gal. (19L) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10611294
View Documents
Epredia™
6764254
3.8 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12539996
View Documents
Epredia™
9990244
Reconstituted to 5 Liters (Ea.) This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12580057
View Documents
Epredia™
9990914
60 mL specimen container filled to 30mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15182527
View Documents
Epredia™
9990916
90 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12590057
View Documents
Epredia™
9990918
120mL specimen container filled to 60mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Phosphate-free
  • Use for storage of all tissue types
  • Stores specimens without excessive shrinking or hardening
  • Available premixed, ready-to-use, or in economical concentrate
  • Concentrate is not affected by freezing, reduces shelf space, and is less expensive to ship
  • Pre-filled, leak-resistant cups are available
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1A
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after single exposure Category 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H350-May cause cancer.
  • H370-Causes damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : formaldehyde, methylalcohol, maleic acid, sodium hydroxide

SDS
Documents

Documents

Product Certifications