Learn More
Epredia™ Glyo-Fixx™
Obtain superior results to neutral buffered formalin with Glyo-Fixx™, a noncarcinogenic, nonsensitizing, and environmentally friendly substitute.
Brand: Epredia™ 9990928
154.85 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Produces distinct and crisp cellular morphology
- Fixes most antigens without impairing immunoreactivity
- Allows special stain dyes, including silver procedures, to be sharp without background staining
- Presents fine detail in specimen structure
Specifications
Fixative | |
125 x 120mL |