Learn More
Epredia™ Shandon™ Gram Stain
Differentiate between Gram-positive and Gram-negative bacteria with Epredia™ Shandon™ Gram Stain.
Brand: Epredia™ 9990725
Includes: Gram crystal violet stain, 4L
Description
- Based upon the popular Hucker modification
- Demonstrates the presence of microorganisms such as bacteria
Specifications
Gram Crystal Violet stain | |
Crystal violet gram stain | |
4L |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Shandon Crystal Violet Stain
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol, C.I. basicviolet 1