Learn More
Epredia™ Shandon™ Harris Hematoxylin
Take advantage of acidified or non-acidified formulations with the Epredia™ Shandon™ Harris Hematoxylin. This product does not employ a mercury-based oxidizing agent, thus removing a possible source of environmental contamination.
Brand: Epredia™ 6765004
185.73 GBP valid until 2025-12-31
Use promo code "24374" to get your promotional price.
Description
Acidifed: Intensely stained, blue-black nuclei
Non-acidified: Intensely stained, blue-black nuclei
Specifications
Hematoxylin | |
4L |
Safety and Handling
- shandon harris hematoxylin acidified
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Supplemental information
- MIXTURE LIST-Contains : ethanol, acetic acid, aluminium sulphate, hematoxylin, sodium iodate
Your input is important to us. Please complete this form to provide feedback related to the content on this product.