Learn More
Epredia™ Shandon™ Harris Hematoxylin
Take advantage of acidified or non-acidified formulations with the Epredia™ Shandon™ Harris Hematoxylin. This product does not employ a mercury-based oxidizing agent, thus removing a possible source of environmental contamination.
Brand: Epredia™ 6765001
109.25 GBP valid until 2025-12-31
Use promo code "24374" to get your promotional price.
Description
Acidifed: Intensely stained, blue-black nuclei
Non-acidified: Intensely stained, blue-black nuclei
Specifications
Hematoxylin | |
2 x 1L |
Safety and Handling
- Danger
Hazard Category
- Carcinogenicity Category 1A
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H350-May cause cancer.
- H371-May cause damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : sodium iodate, aluminium ammonium sulphate dodecahydrate, hematoxylin, ethyl alcohol, methyl alcohol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.