missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Harris Hematoxylin

Take advantage of acidified or non-acidified formulations with the Epredia™ Shandon™ Harris Hematoxylin. This product does not employ a mercury-based oxidizing agent, thus removing a possible source of environmental contamination.

Brand:  Epredia™ 6765003

85.46 GBP valid until 2025-12-31
Use promo code "24374" to get your promotional price.



 View more versions of this product

Product Code. 12647926

  • £107.00 / Pack of 2
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Acidifed: Intensely stained, blue-black nuclei

  • Non-acidified: Intensely stained, blue-black nuclei

Specifications

Specifications

Hematoxylin
2 x 1L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H371-May cause damage to organs.

Precautionary Statement
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P362-Take off contaminated clothing.

Supplemental information
  • MIXTURE LIST-Contains : acetic acid, hematoxylin, sodium iodate,methyl alcohol, aluminium ammonium sulphate dodecahydrate,

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.