missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Kwik-Diff™ Stains

Enjoy a quicker alternative to traditional Wright's and Giemsa Staining procedures with Epredia™ Shandon™ Kwik-Diff™ Stains.

Brand:  Epredia™ 9990707

235.20 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.


 View more versions of this product

Product Code. 10549408

  • £324.00 / Each
Estimated Shipment: 28-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Available in kit form or as individual components
  • Fast three-step differential stain for bone marrows, smears, FNAs, and microorganisms such as H. pylori

Includes:

Reagent 3: Methylene Blue
Specifications

Specifications

4 L
Methylene Blue
(4 L) Kwik-Diff Reagent 3, Methylene Blue
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers