missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Kwik-Diff™ Stains

Enjoy a quicker alternative to traditional Wright's and Giemsa Staining procedures with Epredia™ Shandon™ Kwik-Diff™ Stains.

Brand:  Epredia™ 9990705

232.00 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.



 View more versions of this product

Product Code. 10633649

This item is not returnable. View return policy

Description

Description

  • Available in kit form or as individual components
  • Fast three-step differential stain for bone marrows, smears, FNAs, and microorganisms such as H. pylori

Includes:

Reagent 1: Fixative
Specifications

Specifications

4 L
Fixative
(4 L) Kwik-Diff Reagent 1, Fixative
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 1
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H370-Causes damage to organs.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P271-Use only outdoors or in a well-ventilated area.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P235-Keep cool.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P363-Wash contaminated clothing before reuse.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : methyl alcohol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers