missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ Kwik-Diff™ Stains
Enjoy a quicker alternative to traditional Wright's and Giemsa Staining procedures with Epredia™ Shandon™ Kwik-Diff™ Stains.
Brand: Epredia™ 9990706
234.40 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Available in kit form or as individual components
- Fast three-step differential stain for bone marrows, smears, FNAs, and microorganisms such as H. pylori
Includes:
Reagent 2: Eosin
Specifications
4 L | |
Eosin | |
(4 L) Kwik-Diff Reagent 2, Eosin |