missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon-Mount™

Perform quick, high-quality conventional coverslipping with Shandon-Mount™ permanent mounting medium.

Brand:  Epredia™ 1900333

33.14 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.



 View more versions of this product

Product Code. 12825443

  • £40.85 / 120mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Overnight-drying liquid cover slip
  • Toluene base
  • Refractive index: 1.495 ±0.005 (dry)
  • Viscosity: Low (72-120 cps at 22°C)
Specifications

Specifications

Mounting Medium
120 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1A
  • Acute toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H360-May damage fertility or the unborn child.
  • H371-May cause damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P235-Keep cool.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P363-Wash contaminated clothing before reuse.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : toluene, butyl methacrylate, 2,6-ditertbutyl-p-cresol, butylbenzyl phthalate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers