Learn More
Epredia™ Shandon™ Nu-Clear™ Reagent
Achieve improved cell clarity by removing excess hematoxylin from the slide with Epredia™ Shandon™ Nu-Clear™, a specially-formulated acid alcohol solution.
Brand: Epredia™ 6769009
Description
- Removes residual hematoxylin staining the glass.
- Designed to be used on the full line of Epredia Shandon automatic stainers as well as manual methods.
Nu-Clear I
- Fast-acting solution
- Recommended to be used as a 10-second step in most protocols
- Particularly beneficial for use with Harris hematoxylin
- May be used on the Epredia Varistain™ 24-4 or Shandon Varistain Gemini slide stainers
Nu-Clear II
- Less concentrated
- Recommended as a 20-second step
- May be used on the Epredia Shandon Linistat and other linear stainers
- Rcommended for use with Gill hematoxylins

Specifications
Tissue Processing Reagent (Nu-Clear II) |
Safety and Handling
- Danger
Hazard Category
- Carcinogenicity Category 1A
- Flammable liquid Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H350-May cause cancer.
- H370-Causes damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P235-Keep cool.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ehtyl alcohol, methyl alcohol, methylisobutyl ketone, hydrochloric acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.