Learn More
Epredia™ Peel-A-Way™ Disposable Embedding Molds
Promote rapid paraffin hardening with Epredia Peel-A-Way Disposable Embedding Molds.
£126.15 - £317.86
Product Code | Brand | Dimensions (L x W x H) | Shape | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Shape | Price | Quantity & Availability | ||||
12677736
|
Epredia™
1830 |
22 x 30 x 20 mm | Rectangular |
£317.86
Pack of 288 |
Estimated Shipment: 11-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12731548
|
Epredia™
2219 |
22 x 22 x 20 mm | Square |
£281.00
Pack of 288 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12647736
|
Epredia™
1220 |
22 x 22 mm tapered to 12 x 12 mm, 20 mm deep | Truncated, tapered |
£311.00
Pack of 288 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12687736
|
Epredia™
1841 |
22 x 40 x 20 mm | Rectangular |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11919671
|
Epredia™
1220 |
22 x 22 mm tapered to 12 x 12 mm, 20 mm deep | Truncated, tapered |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Constructed of rigid transparent plastic.
Specifications
Plastic |