missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Scalpel Blade No. 20, Sterile, 1.75 in. (4.5cm)

Cut precisely with the Epredia™ Shandon™ Scalpel Blade No. 20, which is 1.75 in. (4.5cm) in length.

Brand:  Epredia™ 53220

Product Code. 15668765

  • £92.15 / Pack of 100
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Quality patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Defect-free in material and workmanship
Specifications

Specifications

# 20
4.5 cm
10/Pk.
1.75 in.
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers