Learn More
Epredia™ Stainless-Steel Embedding Base Molds
Optimize tissue embedding with the superior thermal exchange of our high-quality, durable Stainless-Steel Embedding Base Molds.
Brand: Epredia™ 6401018
164.05 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Sizes fit most cassettes
- Superior thermal exchange
- Rounded corners
Specifications
Base Mold | |
Stainless Steel | |
24 x 37 x 9 mm |