Learn More
Epredia™ Stainless-Steel Embedding Base Molds
Optimize tissue embedding with the superior thermal exchange of our high-quality, durable Stainless-Steel Embedding Base Molds.
Brand:  Epredia™ 6401017
Code : 50
Additional Details : Weight : 0.11340kg
 View more versions of this product
Product Code. 12697906
  • £334.00 / Pack of 10
Estimated Shipment: 11-07-2024
to see stock.
Description
Description
  • Sizes fit most cassettes
  • Superior thermal exchange
  • Rounded corners
Specifications
Specifications
Base Mold
Stainless Steel
24 x 30 x 9 mm