missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ Stainless-Steel Deep Instrument Trays, 18.1x 12.4 x 2.4 in. (L x W x D) (46 x 31.4 x 6cm)
Epredia™ Shandon Stainless-Steel Deep Instrument Trays are available in three sizes.
£170.00
Product Code | Brand | Length (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (English) | Price | Quantity & Availability | |||||
13457509
|
Epredia™
6127 |
18.1 in. | N/A | ||||||
Description
Instrument Tray Sizes:
- 14.9 x 10.2 x 2.1 in. (37.8 x 25.8 x 5.4cm)
- 16.1 x 11.1 x 2.1 in. (40.9 x 28.1 x 5.4cm)
- 18.1 x 12.4 x 2.4 in. (46 x 31.4 x 6cm)
Specifications
Instrument Trays | |
Stainless Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ Shandon™ Stainless-Steel Deep Instrument Trays, 18.1x 12.4 x 2.4 in. (L x W x D) (46 x 31.4 x 6cm)