Learn More
Epredia™ Shandon™ Suresorb™ Clinical Pads and Formaldehyde Neutralizer Pads
Absorb and lock in liquids with Epredia™ Shandon™ Suresorb™ Clinical Pads and Formaldehyde Neutralizer Pads.
Brand:  Epredia™ 56411
 View more versions of this product
Product Code. 12532736
  • £262.00 / Pack of 50
Explore more special offers
Description
Description
  • Fiber resists tearing, even when saturated
  • Formaldehyde neutralizer pads reduce exposure by chemically bonding with formaldehyde, resulting in the encapsulation of vapors—ideal for grossing areas and autopsy work
  • Clinical pads work as surgical cutting board pads, dissecting pads, autopsy table pads and liners for body trays and cadaver bags
Specifications
Specifications
Formaldehyde Neutralizer Pad
10 in.
8 in.
Formaldehyde
50/Cs.
203.2 mm