Learn More
Epredia™ TBD-1™ Decalcifier
Product Code. p-4532362 Shop All Epredia Products
Click to view available options
Quantity:
2 Pk.
4 Pk.
Unit Size:
4 L
Pack of 2
Product Code. 10012872
Brand: Epredia™ 6764001
Process bone marrow biopsies in as little as 15 to 30 minutes with this rapid hydrochloric acid decalcifier.
  • Process thin pieces of dense bone in 3 to 12 hours
  • Permits utilization of routine end point tests, such as calcium oxalate
  • Protects cells from actions of acids with added ingredients
TRUSTED_SUSTAINABILITY
Specifications
Product Type Decalcifying Solution
Quantity 2 Pk.
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P271-Use only outdoors or in a well-ventilated area.
  • P234-Keep only in original container.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P363-Wash contaminated clothing before reuse.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P390-Absorb spillage to prevent material damage.
  • P405-Store locked up.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P406-Store in corrosive resistant/polypropylene container with a resistant inner liner.
  • P402-Store in a dry place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid, polyvinyl pyrrolidone, fluorad