Learn More
Epredia™ Shandon™ TBD-2™ Decalcifier
Process specimens up to 48 hours past end point undamaged with Epredia™ Shandon™ TBD-2™ Decalcifier, a formic acid decalcifier.
Brand: Epredia™ 6764004
Code : LE
Additional Details : CAS Number : 64-18-6 Weight : 4.00000kg Transport : UN number : 3412 Chem class : 8 Pack group : II
Description
- Protects cells from actions of acid with added ingredients
- Permits utilization of routine end point tests, such as calcium oxalate
Specifications
Decalcifying Solution | |
4 Pk. |
Safety and Handling
- Shandon TBD-2 Decalcifier
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H371-May cause damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P235-Keep cool.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P363-Wash contaminated clothing before reuse.
- P370+P378-In case of fire: Use to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.