missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Tissue-Marking Dyes, Tissue marking dye kit

Permanently mark the margins of excised surgical specimens for exact orientation with Epredia™ Shandon™ Tissue-Marking Dyes.

Brand:  Epredia™ 3120124

42.64 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.



 View more versions of this product

Product Code. 10413229

  • £58.60 / Each
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Special binding pigment evenly coats the tissue without penetrating tissue surface
  • Does not interfere with diagnostic interpretation
  • Quick-drying and easy-to-use; decreases time-factor in urgent frozen-sectioning

Ordering Information:

Available individually or as a kit (includes one bottle each of black, blue, yellow, red, and green; 50 applicator sticks; storage base).

Specifications

Specifications

Blue
Tissue Marking Dye
59 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Shandon Blue Tissue Marking Dye

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers