missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Xylene Substitute

Achieve safer, more convenient processing, staining and coverslipping with Epredia™ Xylene Substitute.

Brand:  Epredia™ 6764506

377.15 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.



 View more versions of this product

Product Code. 10524305

  • £433.00 / Pack of 4

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Not sensitizing to normal skin
  • Evaporation rate similar to xylene
  • Virtually odorless aliphatic hydrocarbon—no citrus aroma
  • Airborne exposure limit three times safer than xylene (TLV 300ppm)
Specifications

Specifications

Xylene Substitute
4 x 1 gal.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P262-Do not get in eyes, on skin, or on clothing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

Supplemental information
  • MIXTURE LIST-Contains : naptha (petroleum),hydrotreated heavy

SDS
Documents

Documents

Certificates
Special Offers

Special Offers