Learn More
Sharpsafe™ Sharps Bin, Infectious Sharps for Incineration
Cost-effective solution for the suitable disposal of infectious sharps.
Brand:  Sharpsafe™ 41731430
Product Code. 15820365
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Wide aperture facilitates easy disposal.
  • Translucent lid, clear internal flap and visible fill line help ensure that the bin is not overfilled.
  • In-mold label that cannot be removed.
  • Simple four-click assembly.
  • Large headroom.
  • Inkjet-printed fill line and waste segregation message.
  • Temporary and final closure feature
Specifications
Specifications
0.2 L
Orange
100/Cs.
Disposal of infectious sharps for incineration.