missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hospidex™ Sharpsafe™ Polypropylene Biohazard Container
Use the Sharpsafe™ Polypropylene Biohazard Container to transport dangerous biohazardous goods for disposal.
Brand: Hospidex™ SHA4160
This item is not returnable.
View return policy
Description
- Yellow
- Polypropylene
- NFX 30-505 certification
Specifications
1 L | |
188 mm | |
Autoclavable | |
Polypropylene |
NFX30-500 | |
106 mm | |
For collection of sharps waste treatment activities, pungent, sharp and / or pointed | |
Biohazard Container |