missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sharpsafe™ Polypropylene Biohazard Sharps Container
Use the Sharpsafe™ Polypropylene Biohazard Sharps Container facilitate the safe management of wastes from healthcare activities.
£331.01 - £488.58
Specifications
Color | Yellow |
---|---|
Length (Metric) | 175 mm |
Quantity | 50 Pk. |
Width (Metric) | 175 mm |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11930368
|
Sharpsafe™
SHA4110 |
7 L | N/A | ||||||
11920368
|
Sharpsafe™
SHA4100 |
4 L | N/A | ||||||
Description
- Polypropylene
- 50Pack
- Yellow
Specifications
Yellow | |
175 mm | |
50 Pk. | |
175 mm |