missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Canine IgG2 Heavy Chain Secondary Antibody, FITC, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Bio-Techne NB7335
Additional Details : Weight : 0.00970kg
Description
IgG2 Heavy Chain Polyclonal antibody specifically detects IgG2 Heavy Chain in Canine samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
IgG2 Heavy Chain | |
Polyclonal | |
FITC | |
Constant region of heavy chain of IgG2, immunoglobulin gamma 2 (Gm marker), immunoglobulin heavy constant gamma 2 (Gm marker) | |
Sheep | |
Affinity Purified | |
RUO | |
3501 | |
Canine | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1.0 mg/mL | |
Immunohistochemistry 1:50 - 1:500, Immunocytochemistry/Immunofluorescence 1:50 - 1:500, Immunohistochemistry-Paraffin | |
IGHG2 | |
Dog IgG2 | |
1 mL | |
Secondary | |
By immunoelectrophoresis and ELISA this reacts specifically with dog IgG2. This may cross react with IgG2 from other species. | |
Store at 4C. Do not freeze. |
For Research Use Only