missing translation for 'onlineSavingsMsg'
Learn More

Sheep anti-Rabbit IgG (H+L) Secondary Antibody, HRP, Novus Biologicals™

Sheep Polyclonal Antibody

Brand:  Novus Biologicals NBP1-73253

197.60 GBP valid until 2025-03-28
Use promo code "25306" to get your promotional price.


Product Code. 18505853

  • £247.00 / 2mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgG (H+L)
Polyclonal
HRP
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free
Sheep anti-Rabbit IgG (H+L) Secondary Antibody [HRP] was produced by repeated immunization with rabbit whole IgG molecule in sheep.
2 mg
Adaptive Immunity, Immunology
Reconstitute with 1.0 mL deionized water (or equivalent).
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, ELISA, Immunohistochemistry
LYOPH
Western Blot 1:1000 - 1:5000, ELISA 1:10000 - 1:50000, Immunohistochemistry 1:500 - 1:2500
Sheep
Immunogen affinity purified
RUO
Secondary
Rabbit
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sheep anti-Rabbit IgG (H+L) Secondary Antibody, HRP, Novus Biologicals™ > 2 mg; HRP

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.