missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep IgG Isotype Control, HRP, Novus Biologicals™
Sheep Antibody
Brand: Novus Biologicals NBP1-96876
Additional Details : Weight : 0.00970kg
Description
IgG specifically detects IgG in Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
IgG | |
Lyophilized from 0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with No Preservative | |
IGHG1 | |
Multi-step | |
RUO | |
3500 | |
Reconstitute with 1.0 ml deionized water (or equivalent). | |
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
HRP | |
Immunoglobulin G, ImmunoglobulinG | |
Sheep | |
1 mg | |
Isotype Control (Primary) | |
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti- Sheep IgG and anti-Sheep Serum. | |
Sheep | |
Lyophilized |
For Research Use Only