missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheet Steel Oxygen Cylinder Trolleys
Brand: N/A
Code : 03
Additional Details : Weight : 1.00000kg
Description
The tubular frame has a sheet steel base and retaining straps to hold the cylinder in position. Each trolley is fitted with anti-static metal centred wheels and front 100mm non-marking corner buffers.Specifications
Oxygen Cylinder Trolley | |
460 x 410 x 1100 mm | |
Carrying oxygen cylinders | |
460 mm | |
410 mm |
Sheet Steel | |
Ideal for use in hospitals | |
1100 mm | |
13 kg |