Learn More
Thermo Scientific™ Shelf Kits w/Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers
Maximize storage space with these shelf kits for Thermo Scientific™ Ultra-Low Temperature Freezers.
£620.41 - £1735.00
Product Code | Brand | For Use With (Equipment) | Holds | No. of Racks | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Holds | No. of Racks | Price | Quantity & Availability | |||
12908344
|
Thermo Scientific™
RSK13SD4 |
Forma 900, Forma 7000, TSE and TSD Series (13 cu. ft. models) | Boxes (48/4 Racks) | 4 |
£653.11
Each |
In Stock
Log in to see stock. |
|||
11329351
|
Thermo Scientific™
RSK13SD5 |
HERAfreeze HFU B Series, Revco ExF and DxF Series (13 cu. ft. models) | Boxes (48/4 Racks) | 4 |
£620.41
Each |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||
12918344
|
Thermo Scientific™
RSK17SD4 |
Forma 900, Forma 7000, TSE and TSD Series (17 cu. ft. models) | Boxes (80/4 Racks) | 4 |
£766.69
Each |
Estimated Shipment: 19-02-2025
Log in to see stock. |
|||
11339351
|
Thermo Scientific™
RSK17SD5 |
HERAfreeze HFU B Series, Revco ExF and DxF Series (17 cu. ft. models) | Boxes (80/4 Racks) | 4 |
£765.83
Each |
Estimated Shipment: 18-02-2025
Log in to see stock. |
|||
12928344
|
Thermo Scientific™
RSK23SD4 |
Forma 900, Forma 7000, TSE and TSD Series (23 cu. ft. models) | Boxes (100/5 Racks) | 5 |
£963.75
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
11349351
|
Thermo Scientific™
RSK23SD5 |
HERAfreeze HFU B Series, Revco ExF and DxF Series | Boxes (80/5 Racks) | 5 |
£963.75
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||
11810543
|
Thermo Scientific™
RSK300SD4 |
Thermo Scientific TDE Series (14.9 cu.ft.) | Boxes (75/3 Racks) | 3 |
£638.48
Each |
In Stock
Log in to see stock. |
|||
11359351
|
Thermo Scientific™
RSK300SD5 |
Thermo Scientific TDE Series (14.9 cu.ft.) | Boxes (60/3 Racks) | 3 |
£635.04
Each |
In Stock
Log in to see stock. |
|||
11820543
|
Thermo Scientific™
RSK400SD4 |
Thermo Scientific TSX and TDE Series (19 cu. ft.) | Boxes (100/4 Racks) | 4 |
£989.00
Each |
In Stock
Log in to see stock. |
|||
11369351
|
Thermo Scientific™
RSK400SD5 |
Thermo Scientific TSX and TDE Series (19 cu. ft.) | Boxes (80/4 Racks) | 4 |
£844.14
Each |
In Stock
Log in to see stock. |
|||
11830543
|
Thermo Scientific™
RSK500SD4 |
Thermo Scientific TSX and TDE Series (24.1 cu. ft.) | Boxes (125/5 Racks) | 5 |
£1033.31
Each |
In Stock
Log in to see stock. |
|||
11379351
|
Thermo Scientific™
RSK500SD5 |
Thermo Scientific TSX and TDE Series (24.1 cu. ft.) | Boxes (100/5 Racks) | 5 |
£1058.40
Each |
Estimated Shipment: 04-02-2025
Log in to see stock. |
|||
11840543
|
Thermo Scientific™
RSK600SD4 |
Thermo Scientific TSX and TDE Series (28.8 cu. ft.) | Boxes (150/6 Racks) | 6 |
£1495.00
Each |
In Stock
Log in to see stock. |
|||
11850543
|
Thermo Scientific™
RSK700SD4 |
Thermo Scientific TSX Series (33.5 cu. ft.) | Boxes (175/7 Racks) | 7 |
£1735.00
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
11399351
|
Thermo Scientific™
RSK700SD5 |
Thermo Scientific TSX Series (33.5 cu. ft.) | Boxes (140/7 Racks) | 7 |
£1480.04
Each |
Estimated Shipment: 29-01-2025
Log in to see stock. |
|||
Description
Maximize storage space with Shelf Kits with Racks and Boxes for Thermo Scientific™ Ultra-Low Temperature Freezers, featuring 2 in. boxes with dividers.
Features- Stainless steel racks
- Boxes
- Cell dividers
Specifications
100 |