missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Isotemp™ Shelf
Material: Wire; For Use With: Isotemp gravity convection oven/Incubator, 2.3/2.5 cu. ft.
Brand: Fisherbrand™
Additional Details : Weight : 2.15000kg
Description
- Material: Stainless steel
Specifications
Isotemp gravity convection oven/incubator, 2.3/2.5 cu. ft. | |
Wire | |
Shelf | |
15.20 x 12.28 in. (386 x 312 mm) |