Learn More
Thermo Scientific™ Shelves for Heratherm™ Ovens and Incubators
Perforated and wire mesh shelves
£39.59 - £338.19
Product Code | Brand | Depth (English) | Width (English) | For Use With (Equipment) | Length (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Depth (English) | Width (English) | For Use With (Equipment) | Length (English) | Price | Quantity & Availability | |||
11621571
|
Thermo Scientific™
50125605 |
11.4 in. | 11.4 in. | Heratherm Compact Microbiological Incubator |
£39.59
Each |
In Stock
Log in to see stock. |
||||
10784222
|
Thermo Scientific™
50127771 |
14.8 in. | 14.8 in. | Heratherm General Protocol Microbiological Incubator 100L | 12.9 in. |
£150.03
Each |
In Stock
Log in to see stock. |
|||
10600601
|
Thermo Scientific™
50127772 |
12.9 in. | 12.9 in. | Heratherm General Protocol Microbiological Incubator 180l | 12.9 in. |
£163.58
Each |
In Stock
Log in to see stock. |
|||
10841481
|
Thermo Scientific™
50127770 |
14.8 in. | 14.8 in. | Heratherm General Protocol Microbiological Incubator 60l | 12.9 in. |
£122.06
Each |
In Stock
Log in to see stock. |
|||
10363940
|
Thermo Scientific™
50127774 |
12.9 in. | 12.9 in. | Heratherm Advanced Protocol/Advanced Protocol Security Microbiological Incubator 100l | 17.2 in. |
£150.03
Each |
In Stock
Log in to see stock. ![]() |
|||
10598290
|
Thermo Scientific™
50127773 |
12.9 in. | 12.9 in. | Heratherm Advanced Protocol/Advanced Protocol Security Microbiological Incubator 60l | 12.9 in. |
£122.06
Each |
In Stock
Log in to see stock. |
|||
10160060
|
Thermo Scientific™
50127777 |
19.8 in. | 19.8 in. | Heratherm Advanced Protocol/Advanced Protocol Security Microbiological Incubator 180l | 17.2 in. |
£163.58
Each |
In Stock
Log in to see stock. |
|||
12665316
|
Thermo Scientific™
50135241 |
21.06 in. | 21.06 in. | Heratherm Advanced Protocol Security Incubators 400l imh400-s and General Protocol Incubator 400l | 20.7 in. |
£264.45
Each |
In Stock
Log in to see stock. ![]() |
|||
12675316
|
Thermo Scientific™
50135242 |
21.06 in. | 38.74 in. | Heratherm Advanced Protocol Security Incubators 750l imh750-s and General Protocol Incubator 750l |
£338.19
Each |
In Stock
Log in to see stock. ![]() |
||||
12685316
|
Thermo Scientific™
50135245 |
21.06 in. | 21.06 in. | Heratherm Advanced Protocol Oven 400I OMH 400 and General Protocol Incubator 400l IGS400 |
£230.07
Each |
Estimated Shipment: 01-05-2025
Log in to see stock. |
||||
12605326
|
Thermo Scientific™
50135244 |
20.2 in. | 20.2 in. | Heratherm Advanced Protocol Oven 750I OMH 750 400 V |
£264.45
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||
12695316
|
Thermo Scientific™
50135246 |
21.06 in. | 21.06 in. | Heratherm Advanced Protocol Oven 750l OMH 750 and General Protocol Incubator 750l IGS750 |
£275.03
Each |
Estimated Shipment: 24-04-2025
Log in to see stock. |
||||
Description
Perforated and wire mesh shelves
Specifications
Shelf |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Thermo Scientific™ Shelves for Heratherm™ Ovens and Incubators