Learn More
Dräger™ Short-Term Detector Tubes: Chlorine
Detector tubes
£68.63 - £107.00
Product Code | Brand | Tube Type | Measurement Range | Humidity Range | No. of Strokes | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tube Type | Measurement Range | Humidity Range | No. of Strokes | Price | Quantity & Availability | ||
15778659
|
Dräger™
CH20701 |
Chlorine 50/a | 50 to 500 | Less than40mg/L | 1 |
£86.45
Pack of 10 |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10178231
|
Dräger™
CH24301 |
Chlorine 0.2/a | 0.2 to 30 | Not Applicable | 10/1 |
£107.00
Pack of 10 |
Estimated Shipment: 18-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
10558292
|
Dräger™
6728411 |
Chlorine 0.3/b2 | 0.3 to 5 | Less than15mg/L | 20 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Glass detector tubes are filled with an inert reagent carrier material (silica gel, aluminum oxide, or aluminum silicate) impregnated with an indicating reagent. Reagent produces a colorimetric indication in the presence of a particular gas, vapor, or aerosol. Concentrations can be read directly from the color change on the tube's printed scale. Tubes are hermetically sealed. Most have a two-year shelf life (when stored at room temperature). Available in packs of ten.Note: Some tests require a pretube or comparison tube in addition to the detector tube. These tubes are included in the box of 10, thus reducing the number of detector tubes. The actual number of detector tubes is cited in the ordering charts.
Order Info
Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
Specifications
Chlorine | |
10 |