missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHQ1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27020-1-AP-20UL
This item is not returnable.
View return policy
Description
SHQ1 is the human homolog of yeast Shq1p, a protein that acts as an accessory factor in the assembly of H/ACA ribonucleoproteins. SHQ1 has been shown to bind the major H/ACA core protein and the pseudouridine synthase NAP57, but precedes the assembly role of NAF1 and of the other H/ACA core proteins. SHQ1 assists in the assembly of H/ACA-box ribonucleoproteins that function in the processing of ribosomal RNAs, modification of spliceosomal small nuclear RNAs, and stabilization of telomerase.Specifications
SHQ1 | |
Polyclonal | |
Unconjugated | |
SHQ1 | |
Protein SHQ1 homolog, SHQ1, SHQ1 homolog (S. cerevisiae) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55164 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6PI26 | |
SHQ1 | |
SHQ1 Fusion Protein Ag25559 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |