missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bohlender™ SICCO™ Maxi 1 Desiccators
Designed for storing or drying humidity sensitive products using silica gel. SICCO™ Maxi 1-Desiccators feature a controlled environment, ideal for storing reference materials, retained samples, metallographic specimen, tobacco and DNA-samples.
Brand: Bohlender™ V1852-01
This item is not returnable.
View return policy
Description
- Tight fitting door protects the contents from contamination from the atmosphere
- Reinforced aluminum frame with panels made of acrylic glass
- Includes four shelves made of stainless steel, tray and hygrometer, four casters, usable with up to 17 shelves
- Door with magnetic catch and circular rubber seal
- Four casters (two of the casters with brakes)
- Easy to read electronic hygrometer
- Variable height shelves made of stainless steel
- Telescopic shelves
- Desiccant tray
Specifications
Clear | |
PMMA Acrylic Glass, Aluminum Frame | |
SICCO Maxi 1-Desiccator, PMMA | |
17 | |
580 x 560 x 1150 mm (exterior) | |
540 mm (interior) |
Vertical | |
311L; 30kg per shelf; 160kg total | |
4 | |
-20°C to +70°C | |
945 mm (interior) |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bohlender™ SICCO™ Maxi 1 Desiccators > SICCO Maxi 1-Desiccator, PMMA
Spot an opportunity for improvement?Share a Content Correction