Learn More
Sieve Shaker
For accurate and reproducible sieving
Brand:  Fritsch 03.8020.00
Product Code. 13177913
  • £9540.00 / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Fritsch sieve shakers satisfy all requirements for accurate and reproducible sieving analysis and perform automatic sieve analysis with the control and evaluation software Autosieve. The Analysette 3 Pro guarantees absolutely reproducible sieving results due to automatic amplitude control, programme selection, can be integrated into any quality management system according to DIN EN ISO 9000, and has an RS232 interface.
Sieves, clamping lid, interposed sieve pans and sieve pans with and without outlet are also available for 100mm and 8inch sieves.Test sieves, as well as accessories for micro precision sieving and for use of the Analysette 3 as micro mill (for grinding small sample quantities), are available on request.
For quantitative particle size analysis of dry samples and suspensions of all kinds, by dry and wet sieving with woven test sieves.
Specifications
Specifications
230 V
Sieve Shaker
50 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products