missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIGLEC7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA511679
This item is not returnable.
View return policy
Description
SIGLEC7 Polyclonal Antibody for Western Blot, IHC (P)
Putative adhesion molecule that mediates sialic-acid dependent binding to cells. Preferentially binds to alpha-2,3- and alpha-2,6-linked sialic acid. Also binds disialogangliosides (disialogalactosyl globoside, disialyl lactotetraosylceramide and disialyl GalNAc lactotetraoslylceramide). The sialic acid recognition site may be masked by cis interactions with sialic acids on the same cell surface. In the immune response, may act as an inhibitory receptor upon ligand induced tyrosine phosphorylation by recruiting cytoplasmic phosphatase(s) via their SH2 domain(s) that block signal transduction through dephosphorylation of signaling molecules. Mediates inhibition of natural killer cells cytotoxicity. May play a role in hemopoiesis. Inhibits differentiation of CD34+ cell precursors towards myelomonocytic cell lineage and proliferation of leukemic myeloid cells (in vitro).
Specifications
| SIGLEC7 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9Y286 | |
| SIGLEC7 | |
| KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human SIGLEC7 (D-siglec). | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| SIGLEC7 | |
| Adhesion inhibitory receptor molecule 1; adhesion inhibitory receptor molecule 1, siglec-7; AIRM1; AIRM-1; CD328; CDw328; D-siglec; p75; p75/ AIRM-1; p75/AIRM1; QA79; QA79 membrane protein; sialic acid binding Ig like lectin 7; sialic acid binding Ig-like lectin 7; sialic acid binding immunoglobulin-like lectin 7; sialic acid-binding Ig-like lectin 7; Siglec; SIGLEC19P; SIGLEC7; SIGLEC-7; SIGLECP2 | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 27036 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction