missing translation for 'onlineSavingsMsg'
Learn More

SymLine™ Signal Boxes

Product Code. 15302352
Click to view available options
Length (Metric):
3 m
55 mm
86 mm
Product Type:
Signalbox T1
Signalbox T5
Plug Type:
EU-style
UK-style
US-style
Includes:
1 Sensor Input and 1 Potential-free Output
5 Sensor Inputs and 6 Potential-free Outputs
Alarm at Empty State
Alarm at Full State
T5 with 5 Disc Fill Level Sensors
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15302352

Brand: SymLine™ 108125

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Monitor two filling levels at once. Ideal for integration into lab furniture with SymLine™ Built-in Signal Box.

  • Clever way to change disposal container without interrupting the inflow
  • LEDs and acoustic buzzer warn you before filling level of collection containers reaches critical height
  • When container is changed, alert can be muted via new touchpad

Specifications

Product Type Signalbox T1
For Use With (Equipment) SCAT™ Sensors and Safety Waste Caps
Length (English) 7.1 in.
Length (Metric) 3 m
Plug Type EU-style
Size 180 x 105 x 55 mm
Voltage 220/230 V
For Use With (Application) Monitoring Filling Levels
Includes Alarm at Full State
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.