Learn More
SymLine™ Signal Boxes
Monitor two filling levels at once. Ideal for integration into lab furniture with SymLine™ Built-in Signal Box.
Brand:  SymLine™ 117988
1601.88 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17813530
  • £2200.00 / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Clever way to change disposal container without interrupting the inflow
  • LEDs and acoustic buzzer warn you before filling level of collection containers reaches critical height
  • When container is changed, alert can be muted via new touchpad
Specifications
Specifications
Signalbox T5
3 m
180 x 105 x 55 mm
Monitoring Filling Levels
SCAT Sensors and Safety Waste Caps
EU-style
230 V
T5 with 5 Disc Fill Level Sensors, 5 x 3 m Signal Cables, 5 x 2 m Hook and Loop Fasteners
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SymLine™ Signal Boxes > Signal Boxes; Includes: T5 with 5 Disc Fill Level Sensors, 5 x 3 m Signal Cables, 5 x 2 m Hook and Loop Fasteners; Length (Metric): 3 m; Type: Signalbox T5; Plug Type: EU-style; Voltage: 230 V