missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEPperCHIP™ Signature Microarray
Signature Microarray with 5500 randomized peptides printed in duplicate. The microarray can be used for epitope mapping, serum screening or biomarker discovery.
Brand: PEPperCHIP™ SPC.049.001
This item is not returnable.
View return policy
Description
- Microarray Content: Randomized peptides with the following amino acid frequencies: 6% for R, S, T, W, Y; 5% for A, D, E, F, G, H, I, K, L, M, N, P, Q, V
- Peptide Length: 15 aa random peptides
- Number of Peptides: 5500 printed in duplicate and HA, polio and c-Myc control peptides
Specifications
PEGMA/PMMA | |
1 Slide | |
1 mm | |
Transparent | |
Individual barcode for each microarray | |
High-density Peptide Microarray | |
Non-sterile |
1 array copy per microarray | |
Combinatorial peptide synthesis by amino acid laser printing | |
ISO 9001:2015 | |
RUO | |
Microscopic Glass | |
Antibody validation, epitope mapping, serum screening, biomarker discovery | |
75.4 x 25 mm |