Learn More
Epredia™ Signature Series™ Clarifier™ 1, 2
Product Code. 10603639 Shop All Epredia Products
Click to view available options
Quantity:
1 gal.
4 x 1 gal.
Type:
Clarifier 1
Clarifier 2
Unit Size:
Each
Pack of 4
Product Code. 10603639
Brand: Epredia™ 7402
Enhance the nuclear definition and clarity of Signature Series hematoxylins with Epredia™ Signature Series Clarifier™ 1, 2.
  • Improves contrast between Nuclei and Cytoplasm
  • Increases cell transparency
  • Removes background staining caused by excessive tissue adhesive
  • Removes unwanted hematoxylin staining from adhesive without affecting nuclear staining
Note:
  • Clarifier 1 is recommended with Hematoxylin 7211 and Hematoxylin 1
  • Clarifier 2 is recommended with Hematoxylin 7212 and Hematoxylin 2
TRUSTED_SUSTAINABILITY
Specifications
Type Clarifier 2
Quantity 1 gal.