missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Signature Series™ Clear-Rite 3™
Render complete deparaffinization and clearing during the staining process with Epredia™ Signature Series Clear-Rite 3.
Brand: Epredia™ 6901
Includes: (4) 1-gallon bottles
Description
- Very low odor; Benzene-free
- Use in all tissue processors and automatic stainers
- Does not produce hard or brittle specimens, even after prolonged exposure

Specifications
Clearing Agent | |
4 x 1 gal. |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Flammable liquid Category 2
- Germ cell mutagenicity Category 1B
- Acute toxicity Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H371-May cause damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : proprietary mixtureof aliphatic hydrocarbons
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ Signature Series™ Clear-Rite 3™ > 4 x 1 Gal. (3.79L)
Spot an opportunity for improvement?Share a Content Correction