missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Signature Series™ Flex™ Dehydrating Reagent
Achieve brilliant cytoplasmic staining and complete dehydration with Epredia™ Signature Series Flex™ 100, 95, 80.
Brand: Epredia™ 8101
93.00 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
View more versions of this product
This item is not returnable.
View return policy
Description
- Patented formulation of methyl and isopropyl alcohol minimizes over-dehydration in smaller biopsies
- Excellent for lipid extraction allowing for increased paraffin infiltration
- May be used as a solvent for special stains
Specifications
Flex 100 Dehydrating Reagent | |
1 gal. |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Flammable liquid Category 2
- Specific target organ toxicity Category 1
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H370-Causes damage to organs.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P271-Use only outdoors or in a well-ventilated area.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P235-Keep cool.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P363-Wash contaminated clothing before reuse.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : isopropyl alcohol, methyl alcohol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction