Learn More
Epredia™ Signature Series™ Flex™ Dehydrating Reagent
Achieve brilliant cytoplasmic staining and complete dehydration with Epredia™ Signature Series Flex™ 100, 95, 80.
Brand: Epredia™ 8201
79.20 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
View more versions of this product
This item is not returnable.
View return policy
Description
- Patented formulation of methyl and isopropyl alcohol minimizes over-dehydration in smaller biopsies
- Excellent for lipid extraction allowing for increased paraffin infiltration
- May be used as a solvent for special stains
Specifications
Flex 95 Dehydrating Reagent | |
1 gal. |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Acute toxicity Category 2
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Oxidising liquid Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H370-Causes damage to organs.
- H301-Toxic if swallowed.
- H331-Toxic if inhaled.
- H226-Flammable liquid and vapour.
- H318-Causes serious eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
Supplemental information
- MIXTURE LIST-Contains : isopropyl alcohol, methyl alcohol