missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Signature Series™ Hematoxylin
Yield results which exceed those of Gill or Harris Hematoxylins with this uniquely formulated product.
£162.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10872300
|
Epredia™
7212 |
1 pt. |
£162.00
Pack of 4 |
In Stock
Log in to see stock. |
|||||
Description
- No filtration required
- Stains up to 2500 slides per pint
- Crisp, well-defined nuclear detail
- Slight affinity for acid mucopolysaccharide (mucin) staining
Available in 5 liter packaging
- Reduces plastic by 90%
- Increases stain stability
- Reduces messy stain spills
- Saves 35% storage space versus traditional packaging
Specifications
Hematoxylin |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- STAIN HEMATOXYLIN 7212 4X1PT
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P321-Specific treatment (see on this label).
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Ethylene glycol, Acetic acid
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title