missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Signature Series™ Paraffin

Minimize compression and obtain rapid infiltration with this unique blend of polymers and highly refined paraffin with narrow carbon chain distribution.

Brand:  Epredia™ 8339

 View more versions of this product

Product Code. 12687766

  • £120.00 / Pack of 10
Estimated Shipment: 27-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Triple-filtered to eliminate impurities
  • Type H allows for ultra-thin sections
  • Type L may reduce infiltration times during processing
  • Both Type L and Type H can be used for infiltration and embedding, and have a melting point of 55° to 57°C

Ordering Information: Packaged in easy-to-open, resealable bags.

Specifications

Specifications

54° to 57°C
Paraffin
2 lb. bag (0.9kg)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Hazard Statement
  • EUH210-Safety data sheet available on request.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers