missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ SILBERBRAND Compact Burette Spare Burette Tube

SILBERBRAND Spare Burette Tube is suitable for use with a compact burette.

£23.90 - £24.40

Specifications

Color Clear
For Use With (Equipment) Compact Burette
Material Borosilicate Glass (3.3)
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10560913
View Documents
BRAND™
13933
10 mL
£24.40
Each
In Stock
to see stock.
Add to basket
 
10644943
View Documents
BRAND™
13936
25 mL
£23.90
Each
In Stock
to see stock.
Add to basket
 
Description

Description

  • Constructed of Borosilicate Glass 3.3
  • Clear glass
Specifications

Specifications

Clear
Compact Burette
Borosilicate Glass (3.3)
Videos
SDS
Documents

Documents

Product Certifications