Learn More
Brand™ Silberbrand™ Eterna™ AR-GLAS™ Class B Measuring Pipets
Manufactured from AR-GLAS™ soda lime glass, this Class B Measuring Pipet is ideal in most research, scientific, and other laboratory environments.
Brand:  Brand™ 27069
 View more versions of this product
Product Code. 12055302
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Manufactured from AR-GLAS™ Soda lime glass.
  • Type 3, with zero value at the top.
  • Amber stain marking.
  • Class B.
  • Compliant with DIN EN ISO 835 standards.
  • Calibrated to deliver (TD, Ex).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.01 mL
±0.008 mL
2 x Green
5 mL